Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053572.1 | NP_446024.1 | 15 | 1839 | ||
GenBank mRNA | AB053449.1 | - | 15 | 1839 |