Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053615.1 | NP_446067.1 | 3 | 328 | ||
GenBank mRNA | BC127486.1 | - | 554 | 8364080 | ||
U77582.1 | - | 3 | 328 |