Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cds2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053643.1 | NP_446095.1 | 12 | 1213 | ||
GenBank mRNA | AB052898.1 | - | 12 | 1213 |