Search Thermo Fisher Scientific
Gene Symbol: Pitpnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053742.1 | NP_446194.1 | 3 | 147 | ||
XM_006249522.2 | XP_006249584.1 | 3 | 191 | |||
XM_006249523.2 | XP_006249585.1 | 3 | 191 | |||
GenBank mRNA | BC061538.1 | - | 3 | 179 | 6888321 | |
D21132.1 | - | 3 | 147 |