Gene Symbol: DUSP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007207.5 | NP_009138.1 | 4 | 2236 | ||
NR_111939.1 | 3 | 1244 | ||||
NR_111940.1 | 3 | 1382 | ||||
GenBank mRNA | BC020608.1 | - | 3 | 1243 | ||
BC031405.1 | - | 4 | 2135 | 5182027 | ||
BC063826.1 | - | 4 | 2130 | 6178330 |