Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053731.2 | NP_446183.2 | 5, 6 | 1487 | ||
XM_006246567.2 | XP_006246629.1 | 6, 7 | 1839 | |||
XM_006246568.2 | XP_006246630.1 | 6, 7 | 1673 | |||
GenBank mRNA | AY028417.1 | - | 5, 6 | 1487 |