Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053743.1 | NP_446195.1 | 3 | 529 | ||
GenBank mRNA | AB097113.1 | - | 3 | 442 | ||
BC061720.1 | - | 3 | 498 | 5598479 | ||
D26564.1 | - | 3 | 529 |