Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034015.1 | NP_001029187.1 | 5 | 372 | ||
NM_053816.2 | NP_446268.2 | 5 | 372 | |||
GenBank mRNA | CB791100.1 | - | 5 | 426 | ||
L13040.1 | - | 3 | 152 | |||
L13041.1 | - | 5 | 376 | |||
L14617.1 | - | 4 | 321 | |||
L14618.1 | - | 4 | 321 |
Set Membership: |
G-protein coupled receptor |