Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053844.1 | NP_446296.1 | 1, 2 | 104 | ||
GenBank mRNA | M97255.1 | - | 1, 2 | 104 |