Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040101.1 | NP_001035190.1 | 2, 3 | 551 | ||
NM_001287763.1 | NP_001274692.1 | 3, 4 | 606 | |||
NM_001287764.1 | NP_001274693.1 | 2, 3 | 497 | |||
NM_014392.4 | NP_055207.1 | 2, 3 | 374 | |||
XM_017008022.1 | XP_016863511.1 | 5, 6 | 1791 | |||
GenBank mRNA | AK094573.1 | - | 7, 8 | 2214 | ||
AK125172.1 | - | 3, 4 | 588 | |||
AK299657.1 | - | 2, 3 | 233 | |||
AK312288.1 | - | 2, 3 | 233 | |||
AX775767.1 | - | 2, 3 | 298 | |||
BC001745.1 | - | 2, 3 | 298 | 3510777 | ||
BC034227.1 | - | 5, 6 | 1247 | 5265974 | ||
DA746130.1 | - | 2, 3 | 202 | |||
DA773692.1 | - | 2, 3 | 370 | |||
DC386827.1 | - | 3, 4 | 515 | |||
HY141242.1 | - | 2, 3 | 375 |