Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053896.2 | NP_446348.2 | 2 | 204 | ||
GenBank mRNA | BC098910.1 | - | 2 | 196 | 7455646 | |
BE111477.1 | - | 2 | 204 | |||
U60063.1 | - | 2 | 200 |