Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034022.1 | NP_001029194.1 | 4 | 366 | ||
NM_053814.2 | NP_446266.2 | 4 | 366 | |||
XM_006246422.2 | XP_006246484.1 | 4 | 634 | |||
XM_006246423.2 | XP_006246485.1 | 4 | 634 | |||
XM_006246424.2 | XP_006246486.1 | 4 | 634 | |||
XM_006246425.2 | XP_006246487.1 | 4 | 633 | |||
XM_006246427.2 | XP_006246489.1 | 4 | 634 | |||
XM_006246431.2 | XP_006246493.1 | 4 | 621 | |||
XM_006246432.2 | XP_006246494.1 | 4 | 619 | |||
XM_008767759.1 | XP_008765981.1 | 4 | 634 | |||
XM_008767760.1 | XP_008765982.1 | 4 | 634 | |||
XM_008767761.1 | XP_008765983.1 | 4 | 635 | |||
XM_008767762.1 | XP_008765984.1 | 4 | 621 | |||
GenBank mRNA | AA924848.1 | - | 4 | 366 | ||
AF311311.1 | - | 4 | 296 | |||
BC098911.1 | - | 4 | 352 | 7446349 | ||
FQ222001.1 | - | 4 | 404 |