Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053925.1 | NP_446377.1 | 225 | |||
GenBank mRNA | AB022014.1 | - | 225 | |||
BC158584.1 | - | 235 | 8369120 |