Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053896.2 | NP_446348.2 | 9 | 1019 | ||
GenBank mRNA | BC098910.1 | - | 9 | 1011 | 7455646 | |
U60063.1 | - | 9 | 1015 |