Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053998.2 | NP_446450.2 | 7 | 513 | ||
GenBank mRNA | BC071176.1 | - | 7 | 506 | ||
BC087584.1 | - | 6 | 344 | |||
BC105863.1 | - | 7 | 513 | 7457017 | ||
JN019799.1 | - | 7 | 508 | |||
M83675.1 | - | 5 | 292 |