Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZHX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014943.3 | NP_055758.1 | 3 | 650 | ||
XM_005250836.4 | XP_005250893.1 | 3 | 1155 | |||
XM_005250837.4 | XP_005250894.1 | 4 | 1118 | |||
XM_011516931.2 | XP_011515233.1 | 3 | 483 | |||
XM_011516932.2 | XP_011515234.1 | 4 | 1310 | |||
GenBank mRNA | AB020661.1 | - | 3 | 387 | ||
AB083653.1 | - | 1 | 83 | |||
BC042145.1 | - | 3 | 562 | 5929401 |