Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241570.2 | XP_006241632.2 | 4742 | |||
XM_008765468.1 | XP_008763690.1 | 4793 | ||||
XM_008765469.1 | XP_008763691.1 | 4793 | ||||
GenBank mRNA | X64411.1 | - | 1031 |