Search Thermo Fisher Scientific
Gene Symbol: Akap12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033653.1 | NP_001028825.1 | 2 | 301 | ||
NM_057103.2 | NP_476444.2 | 3 | 530 | |||
XM_006227847.2 | XP_006227909.1 | 2 | 792 | |||
XM_006227848.2 | XP_006227910.1 | 2 | 732 | |||
GenBank mRNA | AY695056.1 | - | 3 | 530 | ||
AY695057.1 | - | 2 | 301 | |||
AY695058.1 | - | 2 | 255 | |||
U41453.1 | - | 3 | 406 |