Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnliprp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057206.1 | NP_476554.1 | 2 | 182 | ||
GenBank mRNA | L09216.1 | - | 2 | 182 |