Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270626.1 | NP_001257555.1 | 10, 11 | 1313 | ||
NM_001270627.1 | NP_001257556.1 | 9, 10 | 1223 | |||
NM_001270628.1 | NP_001257557.1 | 9, 10 | 1307 | |||
NM_057190.2 | NP_476538.2 | 11, 12 | 1382 | |||
XM_006233554.2 | XP_006233616.1 | 10, 11 | 1595 | |||
XM_008761575.1 | XP_008759797.1 | 10, 11 | 1679 | |||
XM_008761576.1 | XP_008759798.1 | 9, 10 | 1589 | |||
GenBank mRNA | AJ293697.1 | - | 11, 12 | 1315 | ||
AJ293698.1 | - | 10, 11 | 1246 | |||
AJ293699.1 | - | 9, 10 | 1156 | |||
AJ534640.1 | - | 9, 10 | 1239 | |||
AJ534641.1 | - | 5, 6 | 377 | |||
AJ534642.1 | - | 10, 11 | 1189 | |||
BC087719.1 | - | 11, 12 | 1343 | 7318793 |