Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GULP1 SIRNA Assays ›
Gene Symbol:
GULP1
Gene Name:
GULP, engulfment adaptor PTB domain containing 1
Gene Aliases:
CED-6, CED6, GULP
Chromosome Location:
Chr.2: 188291669 - 188595926 on Build GRCh38
UniGene ID:
Hs.470887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
5655
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GULP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001252668.1
NP_001239597.1
5
801
NM_016315.3
NP_057399.1
5
801
NR_045562.1
5
753
NR_045563.1
6
1015
XM_006712580.3
XP_006712643.1
5
867
XM_006712581.1
XP_006712644.1
5
761
XM_006712582.1
XP_006712645.1
5
804
XM_006712583.3
XP_006712646.1
6
1083
XM_006712584.3
XP_006712647.1
5
957
XM_006712585.1
XP_006712648.1
4
270
XM_006712589.3
XP_006712652.1
5
761
XM_011511329.1
XP_011509631.1
5
761
XM_011511331.1
XP_011509633.1
3
278
XM_011511332.1
XP_011509634.1
6
872
XM_011511333.1
XP_011509635.1
5
761
XM_011511335.2
XP_011509637.1
5
761
XM_017004303.1
XP_016859792.1
5
761
XM_017004304.1
XP_016859793.1
5
867
XM_017004305.1
XP_016859794.1
5
804
XM_017004306.1
XP_016859795.1
5
1133
XM_017004307.1
XP_016859796.1
5
804
XM_017004308.1
XP_016859797.1
5
761
GenBank mRNA
AF075014.1
-
5
531
AF191771.1
-
5
560
AF200715.1
-
5
566
AK023668.1
-
5
879
AK055718.1
-
5
801
AK314498.1
-
5
595
BC001103.2
-
5
524
3505407
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
lipid transport
phagocytosis, engulfment
apoptotic process
signal transduction
signal transducer activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0