Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzk1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130401.1 | NP_569085.1 | 1, 2 | 179 | ||
XM_006238620.2 | XP_006238682.1 | 2, 3 | 200 | |||
XM_008763968.1 | XP_008762190.1 | 2, 3 | 372 | |||
XM_008763969.1 | XP_008762191.1 | 2, 3 | 200 | |||
XM_008763970.1 | XP_008762192.1 | 2, 3 | 185 | |||
GenBank mRNA | AF110026.1 | - | 1, 2 | 151 | ||
AF402772.1 | - | 1, 2 | 179 |