Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Adcy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130779.2 | NP_570135.2 | 2 | 965 | ||
XM_006239847.2 | XP_006239909.1 | 1 | 701 | |||
XM_006239848.2 | XP_006239910.1 | 1 | 704 | |||
XM_006239849.2 | XP_006239911.1 | 1 | 698 | |||
XM_006239850.2 | XP_006239912.1 | 1 | 707 | |||
XM_008764532.1 | XP_008762754.1 | 2 | 821 | |||
GenBank mRNA | M55075.1 | - | 2 | 890 |