Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130411.2 | NP_569095.1 | 4, 5 | 563 | ||
XM_006230204.2 | XP_006230266.1 | 4, 5 | 647 | |||
GenBank mRNA | AF416730.1 | - | 3, 4 | 459 | ||
AF495469.1 | - | 4, 5 | 563 | |||
BC086971.1 | - | 4, 5 | 539 | 7124429 | ||
FQ225277.1 | - | 1, 2 | 85 | |||
FQ225360.1 | - | 1, 2 | 85 | |||
FQ226379.1 | - | 4, 5 | 551 | |||
FQ226941.1 | - | 4, 5 | 583 | |||
FQ233306.1 | - | 4, 5 | 557 | |||
FQ233390.1 | - | 4, 5 | 559 | |||
FQ233414.1 | - | 4, 5 | 561 | |||
FQ233980.1 | - | 4, 5 | 476 | |||
FQ234750.1 | - | 4, 5 | 561 |