Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130428.1 | NP_569112.1 | 4 | 373 | ||
GenBank mRNA | AB072907.1 | - | 4 | 373 | ||
DQ402976.1 | - | 2 | 182 |