Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LATS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270519.1 | NP_001257448.1 | 4 | 1284 | ||
NM_004690.3 | NP_004681.1 | 4 | 1284 | |||
NR_073033.1 | 5 | 1190 | ||||
XM_006715603.3 | XP_006715666.1 | 4 | 1126 | |||
XM_011536252.2 | XP_011534554.1 | 4 | 1126 | |||
XM_017011474.1 | XP_016866963.1 | 4 | 1126 | |||
XM_017011475.1 | XP_016866964.1 | 4 | 1046 | |||
XM_017011476.1 | XP_016866965.1 | 5 | 1194 | |||
XM_017011477.1 | XP_016866966.1 | 4 | 1126 | |||
XM_017011478.1 | XP_016866967.1 | 5 | 1203 | |||
XM_017011479.1 | XP_016866968.1 | 4 | 1126 | |||
XM_017011480.1 | XP_016866969.1 | 3 | 823 | |||
XM_017011481.1 | XP_016866970.1 | 3 | 968 | |||
XM_017011482.1 | XP_016866971.1 | 4 | 1126 | |||
GenBank mRNA | AF104413.1 | - | 4 | 1284 | ||
AF164041.1 | - | 3 | 768 | |||
AK310422.1 | - | 5 | 1190 | |||
BC002767.1 | - | 4 | 1142 | 3632571 |