Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130736.1 | NP_570092.1 | 2, 3 | 237 | ||
XM_006237650.2 | XP_006237712.1 | 3, 4 | 291 | |||
GenBank mRNA | AF053317.2 | - | 2, 3 | 237 |