Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130816.1 | NP_570829.1 | 4 | 446 | ||
GenBank mRNA | AB079263.1 | - | 4 | 446 |