Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130403.1 | NP_569087.1 | 4 | 359 | ||
GenBank mRNA | AF352572.1 | - | 4 | 359 | ||
AY050673.1 | - | 4 | 359 | |||
FQ230520.1 | - | 4 | 351 |