Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130411.2 | NP_569095.1 | 6 | 806 | ||
XM_006230204.2 | XP_006230266.1 | 6 | 890 | |||
GenBank mRNA | AF416730.1 | - | 5 | 702 | ||
AF495469.1 | - | 6 | 806 | |||
BC086971.1 | - | 6 | 782 | 7124429 | ||
FQ225277.1 | - | 3 | 328 | |||
FQ225360.1 | - | 3 | 328 | |||
FQ225522.1 | - | 2 | 244 | |||
FQ226290.1 | - | 4 | 275 | |||
FQ226941.1 | - | 6 | 826 | |||
FQ233306.1 | - | 6 | 800 | |||
FQ233414.1 | - | 6 | 804 |