Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acaa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130433.1 | NP_569117.1 | 8 | 1108 | ||
GenBank mRNA | FQ216726.1 | - | 8 | 971 | ||
FQ228855.1 | - | 8 | 972 | |||
X05341.1 | - | 8 | 1108 |