Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LPHN3 SIRNA Assays ›
Gene Symbol:
Lphn3
Gene Name:
latrophilin 3
Gene Aliases:
Cirl3, Lec3
Chromosome Location:
Chr.14: 28362176 - 28856419 on Build Rnor_6.0
UniGene ID:
Rn.203386
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
56816
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Lphn3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_130822.1
NP_570835.1
13
2432
XM_008769968.1
XP_008768190.1
13
2536
XM_008769969.1
XP_008768191.1
13
2521
XM_008769970.1
XP_008768192.1
13
2536
XM_008769971.1
XP_008768193.1
13
2536
XM_008769972.1
XP_008768194.1
12
2497
XM_008769973.1
XP_008768195.1
13
2536
XM_008769974.1
XP_008768196.1
13
2536
XM_008769975.1
XP_008768197.1
13
2536
XM_008769976.1
XP_008768198.1
13
2536
XM_008769977.1
XP_008768199.1
12
2332
XM_008769978.1
XP_008768200.1
12
2497
XM_008769979.1
XP_008768201.1
12
2332
XM_008769980.1
XP_008768202.1
13
2536
XM_008769981.1
XP_008768203.1
13
2536
XM_008769982.1
XP_008768204.1
13
2535
XM_008769983.1
XP_008768205.1
13
2535
XM_008769984.1
XP_008768206.1
13
2535
XM_008769985.1
XP_008768207.1
12
2331
XM_008769986.1
XP_008768208.1
12
2331
GenBank mRNA
AF063103.1
-
13
2432
AF081154.1
-
12
2253
AF081155.1
-
12
2253
AF081156.1
-
12
2253
AF081157.1
-
13
2457
AF081158.1
-
13
2457
AF081159.1
-
13
2457
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_198702, BC094668, AK051766, AK147374, BC058992, and others.
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
brain development
G-protein coupled receptor activity
carbohydrate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline