Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRINP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014618.2 | NP_055433.2 | 2 | 619 | ||
GenBank mRNA | AF027734.1 | - | 2 | 581 | ||
AK293649.1 | - | 2 | 593 | |||
BC021560.1 | - | 2 | 583 | |||
BC065196.1 | - | 2 | 572 | 5245480 |