Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST18 SIRNA Assays ›
Gene Symbol:
ST18
Gene Name:
ST18, C2H2C-type zinc finger
Gene Aliases:
NZF3, ZC2H2C3, ZC2HC10, ZNF387
Chromosome Location:
Chr.8: 52110832 - 52409879 on Build GRCh38
UniGene ID:
Hs.655499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
5701
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST18
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014682.2
NP_055497.1
9
989
XM_006716487.1
XP_006716550.1
9
756
XM_011517629.1
XP_011515931.1
10
912
XM_011517631.1
XP_011515933.1
11
953
XM_011517632.1
XP_011515934.1
9
767
XM_011517633.1
XP_011515935.1
9
760
XM_011517634.1
XP_011515936.1
9
956
XM_011517635.1
XP_011515937.1
9
960
XM_011517636.2
XP_011515938.1
8
1037
XM_011517637.1
XP_011515939.1
7
494
XM_011517638.2
XP_011515940.1
6
535
XM_011517641.1
XP_011515943.1
11
1000
XM_011517642.1
XP_011515944.1
10
912
XM_017014047.1
XP_016869536.1
9
1116
XM_017014048.1
XP_016869537.1
9
1120
XM_017014049.1
XP_016869538.1
10
1166
XM_017014050.1
XP_016869539.1
9
953
XM_017014051.1
XP_016869540.1
8
960
XM_017014052.1
XP_016869541.1
10
1170
XM_017014053.1
XP_016869542.1
10
1157
XM_017014054.1
XP_016869543.1
8
1206
XM_017014055.1
XP_016869544.1
8
797
XM_017014056.1
XP_016869545.1
11
1207
XM_017014057.1
XP_016869546.1
9
1010
XM_017014058.1
XP_016869547.1
9
771
XM_017014059.1
XP_016869548.1
9
957
XM_017014060.1
XP_016869549.1
10
994
XM_017014061.1
XP_016869550.1
8
1129
XM_017014062.1
XP_016869551.1
7
881
XM_017014063.1
XP_016869552.1
8
1427
XM_017014064.1
XP_016869553.1
8
1041
XM_017014065.1
XP_016869554.1
10
1161
XM_017014066.1
XP_016869555.1
8
800
XM_017014067.1
XP_016869556.1
8
1210
XM_017014068.1
XP_016869557.1
9
1001
XM_017014069.1
XP_016869558.1
6
698
XM_017014070.1
XP_016869559.1
5
619
XM_017014071.1
XP_016869560.1
6
867
XM_017014072.1
XP_016869561.1
9
1118
XM_017014073.1
XP_016869562.1
9
756
XM_017014074.1
XP_016869563.1
8
1039
XM_017014075.1
XP_016869564.1
10
1203
XM_017014076.1
XP_016869565.1
10
1207
XM_017014077.1
XP_016869566.1
10
844
XM_017014078.1
XP_016869567.1
9
1124
XM_017014079.1
XP_016869568.1
9
1515
XM_017014080.1
XP_016869569.1
7
756
XM_017014081.1
XP_016869570.1
9
1123
XM_017014082.1
XP_016869571.1
9
958
XM_017014083.1
XP_016869572.1
8
798
XM_017014084.1
XP_016869573.1
6
536
GenBank mRNA
AB011107.1
-
9
853
BC031075.1
-
9
886
BC117147.1
-
4
246
40125698
BC117149.1
-
4
246
40125700
BC143872.1
-
4
245
BC143873.1
-
5
334
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f4tdd:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0