Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133299.2 | NP_579833.1 | 2, 3 | 361 | ||
XM_008767180.1 | XP_008765402.1 | 2, 3 | 367 | |||
GenBank mRNA | AF021854.1 | - | 2, 3 | 362 | ||
AF099742.1 | - | 2, 3 | 281 | |||
BC060546.1 | - | 2, 3 | 355 | 6889831 |