Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014779.3 | NP_055594.1 | 1, 2 | 2982 | ||
NR_130136.1 | 1, 2 | 2982 | ||||
XM_011513337.2 | XP_011511639.1 | 1, 2 | 3049 | |||
GenBank mRNA | AB014569.1 | - | 1, 2 | 2982 | ||
AF201290.1 | - | 1, 2 | 338 | |||
AF201292.1 | - | 1, 2 | 138 | |||
BC044643.1 | - | 1, 2 | 2316 | 5454441 |