Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133594.2 | NP_598278.1 | 2 | 275 | ||
XM_008768397.1 | XP_008766619.1 | 3 | 415 | |||
GenBank mRNA | BC058446.1 | - | 238 | 6921523 | ||
BC078746.1 | - | 2 | 209 | 7125132 | ||
CF976332.1 | - | 279 | ||||
FQ221996.1 | - | 242 | ||||
FQ225379.1 | - | 217 | ||||
FQ225921.1 | - | 233 | ||||
FQ225989.1 | - | 233 | ||||
FQ230134.1 | - | 244 | ||||
FQ235035.1 | - | 2 | 156 | |||
L79949.1 | - | 2 | 222 |
Gene Symbol: Sumo2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729458.4 | XP_002729504.1 | 1 | 212 |