Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133611.2 | NP_598295.2 | 2, 3 | 759 | ||
GenBank mRNA | AJ315643.1 | - | 2, 3 | 767 |