Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133548.1 | NP_598232.1 | 3 | 207 | ||
XM_006244289.2 | XP_006244351.1 | 3 | 326 | |||
XM_006244290.2 | XP_006244352.1 | 3 | 327 | |||
XM_006244291.2 | XP_006244353.1 | 3 | 326 | |||
GenBank mRNA | AF355102.1 | - | 3 | 207 | ||
BC071178.2 | - | 3 | 270 | 7100147 | ||
BC087695.1 | - | 3 | 255 | 7312553 |