Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134392.2 | NP_599219.2 | 5 | 588 | ||
GenBank mRNA | AF291466.1 | - | 5 | 804 | ||
BC126081.1 | - | 5 | 521 | |||
BC158581.1 | - | 5 | 537 |