Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF063102.1 | - | 1, 2 | 104 | ||
AF081148.1 | - | 2, 3 | 550 | |||
AF081149.1 | - | 2, 3 | 550 | |||
AF081150.1 | - | 2, 3 | 550 | |||
AF081151.1 | - | 2, 3 | 550 | |||
AF081152.1 | - | 2, 3 | 550 | |||
AF081153.1 | - | 2, 3 | 550 | |||
FM039554.1 | - | 1, 2 | 59 |