Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191578.1 | NP_001178507.1 | 7, 8 | 2261 | ||
XM_006242875.2 | XP_006242937.1 | 10, 11 | 2734 | |||
XM_006242881.2 | XP_006242943.1 | 10, 11 | 2730 | |||
XM_008766141.1 | XP_008764363.1 | 10, 11 | 2731 | |||
GenBank mRNA | X74226.1 | - | 3, 4 | 742 |