Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138512.1 | NP_612521.1 | 5 | 815 | ||
GenBank mRNA | EU861213.1 | - | 5 | 811 | ||
FQ218628.1 | - | 5 | 812 | |||
FQ218929.1 | - | 2 | 329 | |||
M58041.1 | - | 5 | 815 | |||
X53477.1 | - | 5 | 796 |