Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Myo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138893.1 | NP_620248.1 | 1 | 174 | ||
XM_008771384.1 | XP_008769606.1 | 2 | 1148 | |||
GenBank mRNA | AF209114.1 | - | 1 | 174 | ||
AY004215.1 | - | 1 | 174 |