Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277278.1 | NP_001264207.1 | 3 | 288 | ||
NM_138896.2 | NP_620251.1 | 3 | 288 | |||
GenBank mRNA | BC074015.1 | - | 3 | 213 | 7105756 | |
CB577390.1 | - | 3 | 288 | |||
D32249.1 | - | 3 | 224 |