Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138839.2 | NP_620194.1 | 3, 4 | 316 | ||
XM_006247078.2 | XP_006247140.1 | 3, 4 | 794 | |||
XM_006247079.2 | XP_006247141.1 | 4, 5 | 338 | |||
GenBank mRNA | AF411216.1 | - | 3, 4 | 262 | ||
BC061721.1 | - | 3, 4 | 316 | 5598561 |