Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037541.1 | NP_001032630.1 | 2 | 172 | ||
NM_138852.1 | NP_620207.1 | 2 | 172 | |||
GenBank mRNA | AY089984.1 | - | 2 | 172 | ||
AY348322.1 | - | 2 | 172 |