Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138870.1 | NP_620225.1 | 2 | 296 | ||
XM_006228036.2 | XP_006228098.1 | 3 | 323 | |||
XM_006228037.2 | XP_006228099.1 | 3 | 323 | |||
GenBank mRNA | AB029495.1 | - | 2 | 296 |