Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXXC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101654.1 | NP_001095124.1 | 9 | 1818 | ||
NM_014593.3 | NP_055408.2 | 9 | 1806 | |||
XM_011525940.2 | XP_011524242.1 | 10 | 1885 | |||
XM_011525941.2 | XP_011524243.1 | 8 | 1563 | |||
XM_017025718.1 | XP_016881207.1 | 10 | 1873 | |||
XM_017025719.1 | XP_016881208.1 | 8 | 1551 | |||
GenBank mRNA | AB031069.1 | - | 9 | 1301 | ||
AB209883.1 | - | 7 | 1321 | |||
AF149758.1 | - | 9 | 1310 | |||
AK314886.1 | - | 9 | 1309 | |||
AL136862.1 | - | 9 | 1129 | |||
BC014940.2 | - | 9 | 1174 | 3922983 | ||
BC015733.1 | - | 9 | 1132 | 4857493 | ||
BX647189.1 | - | 9 | 1806 |