Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138919.2 | NP_620274.1 | 2 | 464 | ||
XM_006244736.2 | XP_006244798.1 | 2 | 417 | |||
XM_006244737.2 | XP_006244799.1 | 2 | 362 | |||
GenBank mRNA | AY017209.1 | - | 2 | 464 | ||
BC061976.1 | - | 2 | 459 | 5623807 | ||
FQ220352.1 | - | 2 | 358 | |||
U96638.1 | - | 1 | 230 |